Assay Details
Target Gene Details
Entrez Gene ID: | 268878 |
Gene Name: | ATPase type 13A5 |
Gene Aliases: |
C630015F21Rik |
Location: |
Chr.16:29231851-29378764 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp13a5 | NM_001284375.1 | 9 | 1052 | NP_001271304.1 |
| NM_175650.4 | 10 | 1187 | NP_783581.2 | |
| AK083102.1 | 10 | 1162 | ||
| AK083121.1 | 10 | 1188 | BAC38770.1 | |
| AK158355.1 | 10 | 1007 | BAE34470.1 | |
| BC115739.1 | 9 | 985 |

Genomic Map