Assay Details
Target Gene Details
Entrez Gene ID: | 107522 |
Gene Name: | endothelin converting enzyme 2 |
Gene Aliases: |
1810009K13Rik, 6330509A19Rik, 9630025D12Rik, BB127715 |
Location: |
Chr.16:20611601-20646480 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ece2 | NM_139293.2 | 6 | 824 | NP_647454.2 |
| NM_177940.1 | 6 | 1044 | NP_808809.1 | |
| NM_177941.1 | 7 | 1131 | NP_808810.1 | |
| NM_177942.1 | 5 | 737 | NP_808811.1 | |
| AF396699.1 | 6 | 765 | AAK83919.1 | |
| AF489569.1 | 6 | 863 | AAO72356.1 | |
| AF489570.1 | 7 | 950 | AAO72357.1 | |
| AF489571.1 | 5 | 737 | AAO72358.1 | |
| AF489572.1 | 6 | 824 | AAO72359.1 | |
| AK031951.1 | 6 | 832 | BAC27618.1 | |
| AK043473.1 | 4 | 487 | ||
| AU080537.1 | ||||
| BC138052.1 | 6 | 731 | ||
| BC138053.1 | 6 | 731 |
More Information
Set Membership: |
|

Genomic Map