Assay Details
Target Gene Details
Entrez Gene ID: | 320355 |
Gene Name: | lipase, member I |
Gene Aliases: |
D930038D03Rik, lpd1 |
Location: |
Chr.16:75540514-75586072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lipi | NM_001252513.1 | NP_001239442.1 | ||
| AK079027.1 | BAC37507.1 | |||
| AK086576.1 |
More Information
Set Membership: |
|

Genomic Map