Assay Details
Target Gene Details
Entrez Gene ID: | 245841 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide H |
Gene Aliases: |
- |
Location: |
Chr.16:20717826-20722272 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2h | NM_145632.2 | NP_663607.1 | ||
| AK146429.1 | BAE27163.1 | |||
| AK161617.1 | BAE36495.1 | |||
| BC002306.1 | AAH02306.1 | |||
| BC092087.1 | AAH92087.1 | |||
| BC125612.1 | ||||
| BC125638.1 |
More Information
Set Membership: |
|

Genomic Map