Assay Details
Target Gene Details
Entrez Gene ID: | 21762 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 2 |
Gene Aliases: |
9430095H01Rik, AA407121, TEG-190, Tex190 |
Location: |
Chr.16:20651652-20663414 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd2 | NM_134101.2 | NP_598862.1 | ||
| NR_027485.1 | ||||
| AK146482.1 | ||||
| AK150374.1 | BAE29506.1 | |||
| AK151588.1 | BAE30528.1 | |||
| AK152007.1 | BAE30871.1 | |||
| AK152970.1 | ||||
| AK159098.1 | BAE34814.1 | |||
| AK159636.1 | BAE35250.1 | |||
| AK159756.1 | BAE35347.1 | |||
| AK159815.1 | BAE35395.1 | |||
| AK166816.1 | BAE39042.1 | |||
| AK167326.1 | BAE39428.1 | |||
| AK167774.1 | BAE39806.1 | |||
| AK167786.1 | ||||
| AK167909.1 | BAE39916.1 | |||
| AK167995.1 | BAE39985.1 | |||
| AK168131.1 | BAE40100.1 | |||
| AK168398.1 | ||||
| AK168543.1 | ||||
| AK169867.1 | ||||
| AK188526.1 | ||||
| AK198142.1 | ||||
| AK204117.1 | ||||
| AK204693.1 | ||||
| AK209008.1 | ||||
| AK211861.1 | ||||
| AK213801.1 | ||||
| BC021508.1 | AAH21508.1 | |||
| BC023049.1 | AAH23049.1 | |||
| BC024830.1 | AAH24830.1 | |||
| BC027388.1 | AAH27388.1 | |||
| BC027767.1 | AAH27767.1 | |||
| BC027822.1 | AAH27822.1 | |||
| BC028851.1 | AAH28851.1 | |||
| BC031128.1 | AAH31128.1 | |||
| X80422.1 |
More Information
Set Membership: |
|

Genomic Map