Assay Details
Target Gene Details
Entrez Gene ID: | 20462 |
Gene Name: | transformer 2 beta homolog (Drosophila) |
Gene Aliases: |
5730405G21Rik, D16Ertd266e, SIG-41, Sfrs10, Silg41, TRA2beta |
Location: |
Chr.16:22245741-22266000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tra2b | NM_009186.4 | NP_033212.1 | ||
| AK049573.1 | BAC33819.1 | |||
| AK150007.1 | ||||
| AK152519.1 | ||||
| AK152761.1 | ||||
| AK167387.1 | ||||
| AK167544.1 | ||||
| AK195921.1 | ||||
| AK212179.1 | ||||
| BC061177.1 | AAH61177.1 | |||
| BC086795.1 | AAH86795.1 | |||
| X80232.1 | CAA56518.1 |

Genomic Map