Assay Details
Target Gene Details
Entrez Gene ID: | 66849 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 2 |
Gene Aliases: |
0610025N14Rik, 2310007G06Rik, 4930440J04Rik, 5430408E15Rik, D16Ertd248e, IPP-2 |
Location: |
Chr.16:31251541-31275277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r2 | NM_025800.3 | NP_080076.1 | ||
| AK002671.1 | BAB22274.1 | |||
| AK004320.1 | BAB23261.1 | |||
| AK015349.1 | BAB29808.1 | |||
| AK028603.1 | BAC26028.1 | |||
| AK032462.1 | BAC27882.1 | |||
| AK053657.1 | BAC35465.1 | |||
| AK077194.1 | BAC36673.1 | |||
| AK150281.1 | BAE29437.1 | |||
| AK207871.1 | ||||
| BC069885.1 | AAH69885.1 | |||
| BC069886.1 | AAH69886.1 |
More Information
Set Membership: |
|

Genomic Map