Assay Details
Target Gene Details
Entrez Gene ID: | 19385 |
Gene Name: | RAN binding protein 1 |
Gene Aliases: |
Htf9a |
Location: |
Chr.16:18239979-18248694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ranbp1 | NM_011239.2 | NP_035369.2 | ||
| AK002989.1 | BAB22501.1 | |||
| AK008276.1 | BAB25569.1 | |||
| AK088781.1 | BAC40569.1 | |||
| AK151167.1 | BAE30170.1 | |||
| AK153074.1 | BAE31699.1 | |||
| AK166642.1 | BAE38911.1 | |||
| AK167053.1 | BAE39217.1 | |||
| BC061140.1 | AAH61140.1 | |||
| L25255.1 | AAA16195.1 | |||
| X56045.1 | CAA39516.1 | |||
| X56046.1 | CAA39517.1 |
More Information
Set Membership: |
|

Genomic Map