Assay Details
Target Gene Details
Entrez Gene ID: | 67742 |
Gene Name: | SAM domain, SH3 domain and nuclear localization signals, 1 |
Gene Aliases: |
4930571B16Rik, 930571B16Rik, Hacs1, SLy2 |
Location: |
Chr.16:75858794-75909266 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samsn1 | NM_023380.2 | NP_075869.2 | ||
| AF222928.1 | AAG23356.1 | |||
| AK041340.1 | ||||
| AK041566.1 | BAC30989.1 | |||
| AK053946.1 | BAC35601.1 | |||
| BC052906.1 | AAH52906.1 | |||
| BC064778.1 | AAH64778.1 | |||
| DQ333189.1 | ABC59720.1 |

Genomic Map