Assay Details
Target Gene Details
Entrez Gene ID: | 67742 |
Gene Name: | SAM domain, SH3 domain and nuclear localization signals, 1 |
Gene Aliases: |
4930571B16Rik, 930571B16Rik, Hacs1, SLy2 |
Location: |
Chr.16:75858794-75909266 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samsn1 | NM_023380.2 | 5 | 566 | NP_075869.2 |
| AF222928.1 | 5 | 522 | AAG23356.1 | |
| AK041340.1 | 5 | 498 | ||
| AK053946.1 | 5 | 541 | BAC35601.1 | |
| BC052906.1 | 6 | 578 | AAH52906.1 | |
| BC064778.1 | 5 | 566 | AAH64778.1 | |
| DQ333189.1 | 5 | 566 | ABC59720.1 |

Genomic Map