Assay Details
Target Gene Details
Entrez Gene ID: | 20465 |
Gene Name: | single-minded homolog 2 (Drosophila) |
Gene Aliases: |
bHLHe15 |
Location: |
Chr.16:94085198-94127253 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sim2 | NM_011377.2 | 6 | 1239 | NP_035507.2 |
| D63383.1 | 6 | 1245 | BAA09700.1 | |
| D64135.1 | 6 | 1186 | BAA11013.1 | |
| U40576.1 | 6 | 1571 | AAA91202.1 | |
| U42554.1 | 6 | 911 | AAB19098.1 |
More Information
Set Membership: |
|

Genomic Map