Assay Details
Target Gene Details
Entrez Gene ID: | 72020 |
Gene Name: | zinc finger protein 654 |
Gene Aliases: |
1600021C16Rik, 1810008K20Rik, Znf654 |
Location: |
Chr.16:64780347-64851905 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp654 |

Genomic Map