Assay Details
Target Gene Details
Entrez Gene ID: | 77446 |
Gene Name: | HEG homolog 1 (zebrafish) |
Gene Aliases: |
4632417D23Rik, 5530401I02Rik, 9530025L16Rik, BC038614, Gm629, OTTMUSG00000027315 |
Location: |
Chr.16:33684430-33771576 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Heg1 | NM_175256.5 | NP_780465.4 | ||
| BC076596.1 | AAH76596.1 |
More Information
Set Membership: |
|

Genomic Map