Assay Details
Target Gene Details
Entrez Gene ID: | 15205 |
Gene Name: | hairy and enhancer of split 1 (Drosophila) |
Gene Aliases: |
Hry, bHLHb39 |
Location: |
Chr.16:30065357-30067796 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hes1 | NM_008235.2 | NP_032261.1 | ||
| AK132536.1 | BAE21226.1 | |||
| AK133538.1 | BAE21713.1 | |||
| BC018375.1 | AAH18375.1 | |||
| BC051428.1 | AAH51428.1 |
More Information
Set Membership: |
|

Genomic Map