Assay Details
Target Gene Details
Entrez Gene ID: | 19090 |
Gene Name: | protein kinase, DNA activated, catalytic polypeptide |
Gene Aliases: |
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip |
Location: |
Chr.16:15637444-15842252 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkdc | NM_011159.2 | NP_035289.2 | ||
| AB007544.1 | BAA28873.1 | |||
| AB011543.1 | BAA28875.1 | |||
| AK084827.1 | BAE43387.1 | |||
| D87521.1 | BAA19566.1 |
More Information
Set Membership: |
|

Genomic Map