Assay Details
Target Gene Details
Entrez Gene ID: | 67374 |
Gene Name: | junction adhesion molecule 2 |
Gene Aliases: |
1110002N23Rik, 2410030G21Rik, 2410167M24Rik, JAM-2, JAM-B, Jcam2, VE-JAM |
Location: |
Chr.16:84774123-84826380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jam2 | NM_023844.5 | NP_076333.3 | ||
| AF255911.1 | AAF81224.1 | |||
| AJ291757.1 | CAC20699.1 | |||
| AK010616.1 | BAB27064.1 | |||
| AK010826.1 | BAB27208.1 | |||
| AK013914.1 | BAB29053.1 | |||
| AK028757.1 | BAC26102.1 | |||
| AK045095.1 | BAC32219.1 | |||
| AK078128.1 | BAC37139.1 | |||
| AK153636.1 | ||||
| BC028778.1 | AAH28778.1 |
More Information
Set Membership: |
|

Genomic Map