Assay Details
Target Gene Details
Entrez Gene ID: | 27426 |
Gene Name: | N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase |
Gene Aliases: |
AI596180, UCE |
Location: |
Chr.16:5195280-5204040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nagpa | NM_013796.3 | 1 | 56 | NP_038824.2 |
| NR_027804.1 | 1 | 56 | ||
| AK032158.1 | 1 | 57 | BAC27731.1 | |
| AK087468.1 | 1 | 56 | ||
| AK138035.1 | 1 | 49 | BAE23539.1 | |
| AK155450.1 | BAE33269.1 | |||
| AK159629.1 | BAE35243.1 | |||
| AK162633.1 | ||||
| AK168830.1 | 1 | 46 | ||
| AK169539.1 | ||||
| AK170219.1 | BAE41643.1 | |||
| AK187933.1 | ||||
| BC039790.1 | AAH39790.1 |

Genomic Map