Assay Details
Target Gene Details
Entrez Gene ID: | 15260 |
Gene Name: | histone cell cycle regulation defective homolog A (S. cerevisiae) |
Gene Aliases: |
AA138857, D16Ertd95e, Gm15797, N28177, OTTMUSG00000026264, Tuple1 |
Location: |
Chr.16:18876750-18970309 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hira | NM_010435.2 | NP_034565.2 | ||
| AK134646.1 | ||||
| AK135928.1 | BAE22728.1 | |||
| X92590.1 | CAA63334.1 | |||
| X99712.1 | CAA68049.1 |
More Information
Set Membership: |
|

Genomic Map