Assay Details
Target Gene Details
Entrez Gene ID: | 27393 |
Gene Name: | mitochondrial ribosomal protein L39 |
Gene Aliases: |
C21orf8, MRP-L5, ORF22, Rpml5 |
Location: |
Chr.16:84717580-84735302 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl39 | NM_017404.4 | NP_059100.3 | ||
| AF239728.1 | AAF44697.1 | |||
| AK007681.1 | BAB25185.1 | |||
| AK031942.1 | BAC27613.1 | |||
| BC012274.1 | AAH12274.1 | |||
| BC016561.1 | AAH16561.1 |
More Information
Set Membership: |
|

Genomic Map