Assay Details
Target Gene Details
Entrez Gene ID: | 78913 |
Gene Name: | listerin E3 ubiquitin protein ligase 1 |
Gene Aliases: |
4930528H02Rik, AV266914, C87237, Listerin, Rnf160, Zfp294 |
Location: |
Chr.16:87376651-87432619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltn1 | NM_001081068.1 | NP_001074537.1 | ||
| AK019706.2 | ||||
| AK036725.1 | BAC29552.1 | |||
| AK165062.1 | ||||
| AK173009.1 | BAD32287.1 | |||
| BC116272.2 | AAI16273.1 | |||
| BC116273.2 | AAI16274.1 |
More Information
Set Membership: |
|

Genomic Map