Assay Details
Target Gene Details
Entrez Gene ID: | 15260 |
Gene Name: | histone cell cycle regulation defective homolog A (S. cerevisiae) |
Gene Aliases: |
AA138857, D16Ertd95e, Gm15797, N28177, OTTMUSG00000026264, Tuple1 |
Location: |
Chr.16:18876750-18970309 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hira | NM_010435.2 | 7 | 1301 | NP_034565.2 |
| BC052856.1 | 7 | 719 | AAH52856.1 | |
| X75295.1 | 8 | 816 | CAA53043.1 | |
| X92590.1 | 7 | 788 | CAA63334.1 | |
| X99712.1 | 7 | 628 | CAA68049.1 |
More Information
Set Membership: |
|

Genomic Map