Assay Details
Target Gene Details
Entrez Gene ID: | 224105 |
Gene Name: | p21 protein (Cdc42/Rac)-activated kinase 2 |
Gene Aliases: |
5330420P17Rik, A130002K10Rik, AI836325, D16Ertd269e, PAK-2, mKIAA4182 |
Location: |
Chr.16:32016290-32079342 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak2 | NM_177326.3 | NP_796300.1 | ||
| AK037275.1 | ||||
| AK037417.1 | BAC29802.1 | |||
| AK038324.1 | BAC29970.1 | |||
| AK076227.1 | BAC36261.1 | |||
| AK220528.1 | BAD90309.1 | |||
| AY167030.1 | AAN65624.1 | |||
| BC029195.1 | AAH29195.1 | |||
| BC043712.1 | AAH43712.1 | |||
| BC086650.1 | AAH86650.1 |
More Information
Set Membership: |
|

Genomic Map