Assay Details
Target Gene Details
Entrez Gene ID: | 75826 |
Gene Name: | SUMO/sentrin specific peptidase 2 |
Gene Aliases: |
2310007L05Rik, 4930538C18Rik, AI646780, AW554757, Smt3ip2, SuPr-1, mKIAA1331 |
Location: |
Chr.16:22009432-22049269 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Senp2 | NM_029457.3 | NP_083733.1 | ||
| NR_027488.1 | ||||
| AF368904.1 | AAL14437.1 | |||
| AK015987.1 | BAB30067.1 | |||
| AK031030.1 | BAC27222.1 | |||
| AY188288.1 | AAO27902.1 | |||
| BC031652.1 | AAH31652.1 |
More Information
Set Membership: |
|

Genomic Map