Assay Details
Target Gene Details
Entrez Gene ID: | 208169 |
Gene Name: | solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1 |
Gene Aliases: |
Gm610, NHE-10, Slc9a10, sNHE, spermNHE |
Location: |
Chr.16:45535266-45610396 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9c1 | NM_198106.4 | 10 | 1368 | NP_932774.3 |
| AY368685.1 | 10 | 1328 | AAQ88278.1 |
More Information
Set Membership: |
|

Genomic Map