Assay Details
Target Gene Details
Entrez Gene ID: | 26413 |
Gene Name: | mitogen-activated protein kinase 1 |
Gene Aliases: |
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk |
Location: |
Chr.16:16983382-17047453 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk1 | NM_001038663.1 | NP_001033752.1 | ||
| NM_011949.3 | NP_036079.1 | |||
| AK035386.1 | BAC29053.1 | |||
| AK048127.1 | BAC33251.1 | |||
| AK087925.1 | BAC40044.1 | |||
| AK132241.1 | BAE21053.1 | |||
| AK148853.1 | BAE28679.1 | |||
| AK193161.1 | ||||
| BC058258.1 | AAH58258.1 | |||
| D10939.1 | BAA01733.1 | |||
| X58712.1 | CAA41548.1 |
More Information
Set Membership: |
|

Genomic Map