Assay Details
Target Gene Details
Entrez Gene ID: | 26413 |
Gene Name: | mitogen-activated protein kinase 1 |
Gene Aliases: |
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk |
Location: |
Chr.16:16983382-17047453 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk1 | NM_001038663.1 | 8 | 1265 | NP_001033752.1 |
| NM_011949.3 | 8 | 1265 | NP_036079.1 | |
| AK013699.1 | ||||
| AK035386.1 | 8 | 1239 | BAC29053.1 | |
| AK048127.1 | 8 | 1267 | BAC33251.1 | |
| AK087925.1 | 8 | 1240 | BAC40044.1 | |
| AK132241.1 | 8 | 1253 | BAE21053.1 | |
| AK148853.1 | 8 | 1227 | BAE28679.1 | |
| AK211534.1 | 3 | 224 | ||
| BC006708.1 | 3 | 311 | AAH06708.2 | |
| BC058258.1 | 8 | 1208 | AAH58258.1 | |
| D10939.1 | 8 | 1080 | BAA01733.1 | |
| X58712.1 | 8 | 1038 | CAA41548.1 |
More Information
Set Membership: |
|

Genomic Map