Assay Details
Target Gene Details
Entrez Gene ID: | 104015 |
Gene Name: | synaptojanin 1 |
Gene Aliases: |
A930006D20Rik, AA675315, mKIAA0910 |
Location: |
Chr.16:90936092-91012137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj1 | NM_001045515.1 | NP_001038980.1 | ||
| NM_001164483.1 | NP_001157955.1 | |||
| AB093272.1 | BAC41456.2 | |||
| CV558169.1 | ||||
| EL606634.1 |
More Information
Set Membership: |
|

Genomic Map