Assay Details
Target Gene Details
Entrez Gene ID: | 26565 |
Gene Name: | phospholipase A2, group X |
Gene Aliases: |
PLA2GX, mGXsPLA2, sPLA2-X |
Location: |
Chr.16:13715057-13730983 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g10 | NM_001291009.1 | 1 | 471 | NP_001277938.1 |
| NM_011987.3 | 1 | 471 | NP_036117.1 | |
| NR_110990.1 | 1 | 471 | ||
| AF166097.2 | 1 | 73 | AAF04498.2 | |
| CJ047253.1 | 1 | 472 |
Target Gene Details
Entrez Gene ID: | 76684 |
Gene Name: | phospholipase A2, group X, opposite strand |
Gene Aliases: |
1810007I06Rik |
Location: |
Chr.16:13729836-13739471 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g10os | NR_040574.1 | |||
| AK007362.1 |
More Information
Set Membership: |
|

Genomic Map