Assay Details
Target Gene Details
Entrez Gene ID: | 54128 |
Gene Name: | phosphomannomutase 2 |
Gene Aliases: |
AI585868, C86848 |
Location: |
Chr.16:8637707-8657552 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmm2 | NM_016881.2 | NP_058577.1 | ||
| AF043514.1 | AAD02276.1 | |||
| AK002858.1 | BAB22411.1 | |||
| AK003334.1 | BAB22722.1 | |||
| AK005074.1 | BAB23798.1 | |||
| AK012683.1 | BAB28409.1 | |||
| AK145402.1 | BAE26415.1 | |||
| AK149116.1 | BAE28741.1 | |||
| BC046325.1 | AAH46325.1 | |||
| BC094448.1 | AAH94448.1 |
More Information
Set Membership: |
|

Genomic Map