Assay Details
Target Gene Details
Entrez Gene ID: | 19125 |
Gene Name: | proline dehydrogenase |
Gene Aliases: |
Pro-1, Pro1, Ym24d07 |
Location: |
Chr.16:18071726-18089283 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prodh | NM_011172.2 | 4 | 659 | NP_035302.2 |
| AF120279.1 | 4 | 616 | AAD24776.1 | |
| AK144068.1 | 4 | 629 | BAE25683.1 | |
| BB611875.1 | ||||
| BC125327.1 | 4 | 462 | ||
| U80020.1 | 4 | 608 | AAF21467.1 |
More Information
Set Membership: |
|

Genomic Map