Assay Details
Target Gene Details
Entrez Gene ID: | 77626 |
Gene Name: | sphingomyelin phosphodiesterase 4 |
Gene Aliases: |
4122402O22Rik, mKIAA1418 |
Location: |
Chr.16:17619354-17644830 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smpd4 | NM_001164609.1 | 8 | 667 | NP_001158081.1 |
| NM_001164610.1 | 8 | 667 | NP_001158082.1 | |
| NM_001164611.1 | 8 | 667 | NP_001158083.1 | |
| NM_029945.3 | 8 | 667 | NP_084221.2 | |
| AK019466.2 | 8 | 668 | BAB31737.1 | |
| AK031029.1 | 8 | 661 | BAC27221.1 | |
| AK031685.1 | 8 | 666 | BAC27512.1 | |
| AK035892.1 | 8 | 659 | BAC29229.1 | |
| AK036055.1 | 8 | 659 | ||
| AK036380.1 | 8 | 663 | ||
| AK129354.1 | 8 | 644 | BAC98164.1 | |
| AK138720.1 | 8 | 641 | ||
| AK164234.1 | 8 | 659 | BAE37694.1 | |
| AK164456.1 | 6 | 1132 | BAE37796.1 | |
| AK178840.1 | 2 | 216 | ||
| BC026767.1 | 8 | 640 | AAH26767.1 |
More Information
Set Membership: |
|

Genomic Map