Assay Details
Target Gene Details
Entrez Gene ID: | 16155 |
Gene Name: | interleukin 10 receptor, beta |
Gene Aliases: |
6620401D04Rik, AI528744, CRF2-4, Crfb4, D16H21S58, D21S58h, IL-10R2, Il10r2 |
Location: |
Chr.16:91406235-91425834 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il10rb | NM_008349.5 | NP_032375.2 | ||
| AF440787.1 | AAL40946.1 | |||
| AK040938.1 | ||||
| AK134042.1 | BAE21991.1 | |||
| AK160991.1 | BAE36136.1 | |||
| BC145791.1 | ||||
| U53696.1 | AAC53062.1 |
More Information
Set Membership: |
|

Genomic Map