Assay Details
Target Gene Details
Entrez Gene ID: | 83383 |
Gene Name: | transcription factor AP4 |
Gene Aliases: |
AI642933, AP-4, D930048N17Rik, Tcfap4, bHLHc41 |
Location: |
Chr.16:4544661-4560772 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfap4 | NM_031182.2 | 6 | 947 | NP_112459.1 |
| AF161262.1 | 6 | 947 | AAF80448.1 | |
| AK053091.1 | 6 | 739 | BAC35263.1 | |
| AK166424.1 | 6 | 1086 | BAE38767.1 | |
| BC047270.1 | 6 | 854 | AAH47270.1 | |
| BC054777.1 | 6 | 838 | AAH54777.1 |
More Information
Set Membership: |
|

Genomic Map