Assay Details
Target Gene Details
Entrez Gene ID: | 11516 |
Gene Name: | adenylate cyclase activating polypeptide 1 |
Gene Aliases: |
PACAP |
Location: |
Chr.17:93198904-93206541 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcyap1 | NM_009625.2 | NP_033755.1 | ||
| AK079530.1 | BAC37673.1 | |||
| AK134669.1 | BAE22234.1 | |||
| AK142765.1 | ||||
| BC057344.1 | AAH57344.1 | |||
| D14716.1 |
More Information
Set Membership: |
|

Genomic Map