Assay Details
Target Gene Details
Entrez Gene ID: | 11984 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit C |
Gene Aliases: |
Atp6c, Atp6c2, Atp6l, Atpl, Atpl-rs1, PL16, VATL, Vma3 |
Location: |
Chr.17:24163866-24169429 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0c | NM_009729.3 | NP_033859.1 | ||
| AK002570.1 | 1 | 35 | BAB22195.1 | |
| AK002871.1 | BAB22419.1 | |||
| AK014361.1 | 1 | 44 | ||
| AK028239.1 | BAC25834.1 | |||
| AK146159.1 | BAE26942.1 | |||
| AK170346.1 | BAE41735.1 | |||
| AK181498.1 | ||||
| AK187408.1 | ||||
| AK187661.1 | ||||
| AK212163.1 | ||||
| BC050939.1 | AAH50939.1 | |||
| BC083129.1 | ||||
| BC099475.1 | ||||
| M64298.1 | AAA39775.1 |
More Information
Set Membership: |
|

Genomic Map