Assay Details
Target Gene Details
Entrez Gene ID: | 12607 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
AI848081, CBF2, Cbf, Cebpa-rs1 |
Location: |
Chr.17:78919003-78937070 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpz | NM_001024806.2 | 4 | 2056 | NP_001019977.1 |
| AK136173.1 | 4 | 2037 | BAE22857.1 | |
| AK145374.1 | 4 | 2038 | ||
| AK155990.1 | 4 | 2039 | ||
| AK158771.1 | 4 | 2033 | BAE34655.1 | |
| AK159081.1 | 4 | 2053 | BAE34799.1 | |
| CA575354.1 | 2 | 349 | ||
| U19891.1 | 4 | 2027 | AAB01503.1 | |
| U19892.1 | 4 | 1741 | AAB01504.1 |
More Information
Set Membership: |
|

Genomic Map