Assay Details
Target Gene Details
Entrez Gene ID: | 12607 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
AI848081, CBF2, Cbf, Cebpa-rs1 |
Location: |
Chr.17:78919003-78937070 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpz | NM_001024806.2 | NP_001019977.1 | ||
| AK136173.1 | BAE22857.1 | |||
| AK145374.1 | ||||
| AK155990.1 | ||||
| AK158771.1 | BAE34655.1 | |||
| AK159081.1 | BAE34799.1 | |||
| CA575354.1 | ||||
| U19891.1 | AAB01503.1 | |||
| U19892.1 | AAB01504.1 |
More Information
Set Membership: |
|

Genomic Map