Assay Details
Target Gene Details
Entrez Gene ID: | 212517 |
Gene Name: | cilia and flagella associated protein 44 |
Gene Aliases: |
6330444M21Rik, D16Ertd642e, Gm1750, Wdr52 |
Location: |
Chr.16:44394742-44482694 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfap44 | NM_001033247.1 | NP_001028419.1 |

Genomic Map