Assay Details
Target Gene Details
Entrez Gene ID: | 12305 |
Gene Name: | discoidin domain receptor family, member 1 |
Gene Aliases: |
6030432F18, AI323681, CD167a, Cak, Nep, PTK3A |
Location: |
Chr.17:35681567-35704629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddr1 | NM_001198831.1 | NP_001185760.1 | ||
| NM_001198833.1 | NP_001185762.1 | |||
| NM_007584.2 | NP_031610.1 | |||
| NM_172962.1 | NP_766550.1 | |||
| AF026259.1 | AAB81866.1 | |||
| AK031442.1 | BAC27407.1 | |||
| AK138698.1 | ||||
| AK143088.1 | BAE25267.1 | |||
| BC037108.1 | ||||
| BC065998.1 | AAH65998.1 | |||
| L57509.1 | AAB05209.1 |
More Information
Set Membership: |
|

Genomic Map