Assay Details
Target Gene Details
Entrez Gene ID: | 53761 |
Gene Name: | proline-rich coiled-coil 2A |
Gene Aliases: |
3110039B05Rik, Bat-2, Bat2, D17H6S51E, G2, Wbp12, mKIAA4178 |
Location: |
Chr.17:35149076-35165162 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrc2a | NM_001199044.1 | NP_001185973.1 | ||
| NM_020027.3 | NP_064411.2 | |||
| AK220526.1 | BAD90525.1 | |||
| BC006664.1 | AAH06664.1 | |||
| BC053522.1 | AAH53522.1 |
More Information
Set Membership: |
|

Genomic Map