Assay Details
Target Gene Details
Entrez Gene ID: | 14962 |
Gene Name: | complement factor B |
Gene Aliases: |
AI195813, AI255840, B, Bf, C2, Fb, H2-Bf |
Location: |
Chr.17:34856374-34862514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfb | NM_001142706.1 | NP_001136178.1 | ||
| NM_008198.2 | NP_032224.2 | |||
| AK098094.1 | ||||
| AK149531.1 | BAE28943.1 | |||
| AK149573.1 | ||||
| AK151220.1 | BAE30214.1 | |||
| AK151246.1 | BAE30236.1 | |||
| AK151618.1 | BAE30555.1 | |||
| AK151974.1 | BAE30842.1 | |||
| BC005451.1 | AAH05451.1 | |||
| K01496.1 | AAA39549.1 | |||
| K01497.1 | AAA39550.1 | |||
| M57890.1 | AAA63293.1 |

Genomic Map