Assay Details
Target Gene Details
Entrez Gene ID: | 19252 |
Gene Name: | dual specificity phosphatase 1 |
Gene Aliases: |
3CH134, MKP1, Ptpn16, U19515, erp, mkp-1 |
Location: |
Chr.17:26505591-26508472 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp1 | NM_013642.3 | 3 | 772 | NP_038670.1 |
| AK145903.1 | 3 | 777 | ||
| AK149915.1 | 3 | 650 | BAE29163.1 | |
| AK150433.1 | 3 | 772 | BAE29556.1 | |
| AK152183.1 | 3 | 772 | BAE31014.1 | |
| AK152324.1 | 3 | 775 | BAE31126.1 | |
| AK152905.1 | 2 | 1142 | ||
| AK159251.1 | 3 | 773 | BAE34932.1 | |
| AK159363.1 | 3 | 773 | BAE35021.1 | |
| AK159567.1 | 3 | 773 | BAE35190.1 | |
| AK159658.1 | 3 | 770 | BAE35267.1 | |
| AK159726.1 | 3 | 706 | BAE35320.1 | |
| AK167602.1 | 3 | 774 | BAE39659.1 | |
| AK168627.1 | 3 | 773 | BAE40487.1 | |
| BC006967.1 | 3 | 759 | AAH06967.1 | |
| X61940.1 | 3 | 762 | CAA43944.1 |
More Information
Set Membership: |
|

Genomic Map