Assay Details
Target Gene Details
Entrez Gene ID: | 14158 |
Gene Name: | fer (fms/fps related) protein kinase |
Gene Aliases: |
AV082135, C330004K01Rik, Fert, Fert2 |
Location: |
Chr.17:63829950-64146519 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fer | NM_001037997.3 | NP_001033086.2 | ||
| NM_001286415.1 | NP_001273344.1 | |||
| NM_008000.2 | NP_032026.2 | |||
| AK082799.1 | BAC38626.1 | |||
| AK157828.1 | BAE34213.1 | |||
| BC051249.1 | AAH51249.1 | |||
| BC058100.1 | AAH58100.1 | |||
| M32054.1 | AAA37617.1 | |||
| U76762.1 | AAB18988.1 |

Genomic Map