Assay Details
Target Gene Details
Entrez Gene ID: | 13733 |
Gene Name: | adhesion G protein-coupled receptor E1 |
Gene Aliases: |
DD7A5-7, EGF-TM7, Emr1, F4/80, Gpf480, Ly71, TM7LN3 |
Location: |
Chr.17:57358651-57483529 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgre1 | NM_010130.4 | NP_034260.1 | ||
| AK133202.1 | BAE21555.1 | |||
| AK149898.1 | ||||
| AK151303.1 | ||||
| AK151682.1 | BAE30606.1 | |||
| AK155862.1 | BAE33466.1 | |||
| BC075688.1 | AAH75688.1 | |||
| BY197649.1 | ||||
| U66888.1 | AAC53184.1 | |||
| U66889.1 | AAC53185.1 | |||
| U66890.1 | AAC53186.1 | |||
| U66891.1 | AAC53187.1 | |||
| U66892.1 | AAC53188.1 | |||
| X93328.1 | CAA63720.1 |
More Information
Set Membership: |
|

Genomic Map