Assay Details
Target Gene Details
Entrez Gene ID: | 252870 |
Gene Name: | ubiquitin specific peptidase 7 |
Gene Aliases: |
2210010O09Rik, AA409944, AA617399, AU019296, AW548146, C80752, Hausp |
Location: |
Chr.16:8688722-8757067 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp7 | NM_001003918.2 | 28 | 3035 | NP_001003918.2 |
| AF548565.1 | 28 | 2978 | AAQ12339.1 | |
| AK075830.1 | 6 | 487 | BAC35992.1 | |
| BC100666.1 | 23 | 2350 | AAI00667.1 |
More Information
Set Membership: |
|

Genomic Map