Assay Details
Target Gene Details
Entrez Gene ID: | 271377 |
Gene Name: | zinc finger and BTB domain containing 11 |
Gene Aliases: |
9230110G02Rik, ZNF-U69274 |
Location: |
Chr.16:55972697-56008913 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb11 | NM_173026.2 | 8 | 2498 | NP_766614.2 |
| AK033794.1 | 5 | 1179 | BAC28477.1 |
More Information
Set Membership: |
|

Genomic Map