Assay Details
Target Gene Details
Entrez Gene ID: | 72886 |
Gene Name: | coiled-coil domain containing 94 |
Gene Aliases: |
2900016D05Rik, AI413813 |
Location: |
Chr.17:55959187-55968285 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc94 | NM_028381.3 | NP_082657.1 | ||
| AK013533.1 | BAB28900.1 | |||
| AK168621.1 | ||||
| BC031400.1 | AAH31400.1 |
More Information
Set Membership: |
|

Genomic Map