Assay Details
Target Gene Details
Entrez Gene ID: | 52614 |
Gene Name: | adhesion G protein-coupled receptor E4 |
Gene Aliases: |
D17Ertd479e, Egf-tm7, Emr4, Fire, Gpr127 |
Location: |
Chr.17:55749984-55853662 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgre4 | NM_139138.3 | 9 | 935 | NP_631877.2 |
| AF396935.1 | 9 | 893 | AAL31879.1 | |
| AK036459.1 | 9 | 937 | ||
| AK037483.1 | 9 | 880 | BAC29816.1 | |
| AK155935.1 | 9 | 883 | BAE33510.1 | |
| AY032690.1 | 9 | 854 | AAK51125.1 | |
| BC120538.1 | 9 | 892 | ||
| BC120540.1 | 9 | 892 |
More Information
Set Membership: |
|

Genomic Map