Assay Details
Target Gene Details
Entrez Gene ID: | 224065 |
Gene Name: | urotensin 2B |
Gene Aliases: |
Gm538, U2B, Urp, Uts2d |
Location: |
Chr.16:27353322-27370239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uts2b | NM_198166.3 | NP_937809.1 | ||
| AB116020.1 | BAC98928.1 | |||
| AK131972.1 | BAE20912.1 | |||
| AK131973.1 | BAE20913.1 | |||
| AK132025.1 | BAE20950.1 | |||
| AV144685.2 | ||||
| AY321315.1 | AAQ83885.1 | |||
| BB611261.1 | ||||
| BC117024.1 | ||||
| BC117026.1 |

Genomic Map