Assay Details
Target Gene Details
Entrez Gene ID: | 55938 |
Gene Name: | apolipoprotein M |
Gene Aliases: |
1190010O19Rik, G3a, NG20 |
Location: |
Chr.17:35128997-35131752 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apom | NM_018816.1 | 6 | 645 | NP_061286.1 |
| AF207820.1 | 6 | 645 | AAF23407.1 | |
| AK004530.1 | 6 | 690 | BAB23349.1 | |
| BC021597.1 | 6 | 682 | AAH21597.1 |
More Information
Set Membership: |
|

Genomic Map